Skip to Content
Merck
All Photos(1)

Key Documents

808741

Sigma-Aldrich

Isopropyl methanesulfonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O3S
CAS Number:
Molecular Weight:
138.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

Fp: 219°F

Assay

95%

form

liquid

refractive index

n20/D 1.419

density

1.1351 g/mL at 25 °C

SMILES string

[S](=O)(=O)(OC(C)C)C

InChI

1S/C4H10O3S/c1-4(2)7-8(3,5)6/h4H,1-3H3

InChI key

SWWHCQCMVCPLEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F

Flash Point(C)

104 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Marchetti et al.
Archives of toxicology, 95(3), 1103-1116 (2021-01-29)
The Organisation for Economic Co-Operation and Development Test Guideline 488 (TG 488) uses transgenic rodent models to generate in vivo mutagenesis data for regulatory submission. The recommended design in TG 488, 28 consecutive daily exposures with tissue sampling three days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service