主要文件
Z743535
Richter Anti-Vibration pads
L × W 7.0 in. × 8.5 in., pkg of 2
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.35
包裝
pkg of 2
製造商/商標名
Diversified Biotech VIBE-1000
長度 × 寬度
7.0 in. × 8.5 in.
正在寻找类似产品? 访问 产品对比指南
一般說明
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
历史批次信息供参考:
分析证书(COA)