Key Documents
Z742625
Tube-O-DIALYZER Floats
for Micro Tube-O-DIALYZER
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41123100
NACRES:
NB.24
特點
for Micro Tube-O-DIALYZER
包裝
pkg of 6 ea
製造商/商標名
G-BioSciences 786-141F
正在寻找类似产品? 访问 产品对比指南
一般說明
Designed for dialysis in Tube-O-Tanks or a beaker
Replacement ”Tube-O-DIALYZER” floats
法律資訊
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.