主要文件
Z332194
Bottle adapters for BRAND® Dispensers and Burettes
GL 22 thread, GL 22 bottle, polypropylene
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.01
材料
polypropylene
製造商/商標名
BRAND 704322
接頭
GL 22 bottle
GL 22 thread
正在寻找类似产品? 访问 产品对比指南
一般說明
For all Dispensette® models as well as seripettor® and QuikSip
其他說明
BR704322 (India)
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)