主要文件
Z325295
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 1 mL, two marks
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
材料
AR-Glas®
描述
partial delivery
特點
class AS
包裝
pack of 12 ea
製造商/商標名
BRAND 29722
體積測量範圍
1 mL, accuracy: 0.008 mL
容量
1 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
其他說明
BR29722 (India)
法律資訊
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)