主要文件
OH30087569
Ohaus® pH Electrode
STMICRO8
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115604
包裝
pack of 1 ea
製造商/商標名
Ohaus® 30087569
pH測量範圍
0.00-14 pH unit
正在寻找类似产品? 访问 产品对比指南
一般說明
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
應用
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
特點和優勢
Can be used in conjunction with all Starter bench and portable meters
法律資訊
Ohaus is a registered trademark of Ohaus Corp.
从最新的版本中选择一种:
分析证书(COA)