Merck
所有图片(4)
主要文件
HS120018
Work 2 Store® Expanding Tube Storage Box
white, holds 64 x 1.5/2.0 mL Tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122800
NACRES:
NB.35
材料
polycarbonate
polypropylene
white
特點
holds 64 x 1.5/2.0 mL Tubes
包裝
pack of 1 ea
製造商/商標名
Heathrow Scientific HS120018
長度 × 寬度 × 高度
310 mm × 128 mm × 34 mm
一般說明
The Work 2 Store is an expandable single rack and storage box in one. Reduce sample handling time and the need for several racks when working with samples for cold temperature storage. The opening and closing mechanism allows the rack to expand, creating needed room for sample manipulation. When done, simply collapse the rack back to its compact form and it′s ready for the freezer. The Work 2 Store holds 1.5/2.0 mL cryo and microtubes.
特點和優勢
  • Expanding wells have a center depression to stabilize conical tubes and a cryogenic vial lock for one handed cap removal
  • Holds 64 x 1.5/2.0 mL cryo and/or microtubes
  • Sides made from polycarbonate, Tube holders made of polypropylene
  • Instantly creates more working space
  • Helps reduce potential cross contamination
  • Stops opened microfuge lids from interfering with other wells
  • Stackable
  • Alphanumeric identification for quick location and retrieval of tubes
  • Clear lid locks the rack together for safe handling and transportation
  • Will fit into standard 2" (53mm) freezer racking
  • Temperature range -80 to 121ºC. Must use tubes capable of being stored at the required temperature of the user
  • Autoclavable
法律資訊
Work 2 Store is a registered trademark of Heathrow Scientific LLC
历史批次信息供参考:
分析证书(COA)