主要文件
G6276
Grid storage box
plastic (anti-static)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41123300
NACRES:
MA.01
材料
plastic (anti-static)
包裝
pack of 10 ea
製造商/商標名
Ted Pella SB100
相关类别
一般說明
Indexed card holds 100, 2.3 mm or 3.05 mm grids in coded locations. Accompanying record card is used to record identity of samples. Device is anti-static to prevent handling problems due to repulsion or attraction.
历史批次信息供参考:
分析证书(COA)