主要文件
DWP961000SY20-EP
Eppendorf® Deepwell Plate 96/1000μL
sterile, volume 1000 μL, yellow border with clear wells, pkg of 20 plates (5 bags x 4 plates)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122107
尺寸:
96 well
容積:
1000 μL
:
96
無菌:
sterile
特點:
yellow border with clear wells
無菌
sterile
特點
yellow border with clear wells
包裝
pkg of 20 plates (5 bags x 4 plates)
製造商/商標名
Eppendorf® 951032727
存貨情形
available only in North America
技術
PCR: suitable
尺寸
96 well
容積
1000 μL
96
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Eppendorf® Deepwell Plates 96 and 384 are high-performance plates for all manual and automated applications – from sample storage at -86 ºC to DNA denaturation at 100 ºC. Manufactured from pure polypropylene (PP), these plates offer innovative features: More dependability. More efficiency. More stability. The plates are available with a white, yellow, green, or blue border. For easy identification of wells, a high-contrast alphanumeric label is laser-engraved in the border. This OptiTrack® matrix allows you to find your sample up to 30 % more quickly while reducing pipetting errors. With all of these sophisticated features, these plates set new standards in your laboratory.
特點和優勢
  • OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • Minimal remaining volume and high uniformity from well to well
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
  • Autoclavable (121?°C, 20?min)
  • Available with barcode. Additional details available at: www.eppendorf.com/barcode
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
从最新的版本中选择一种:
分析证书(COA)