Key Documents
AXYSCT200SSRS
Self standing screw cap tubes and attached caps with O-rings
capacity 2.0 mL, red, sterile
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.22
材料
red
screw top cap
無菌
sterile
特點
RCF 20,000 × g
包裝
pack of 500 tubes
case of 8 packs
製造商/商標名
Axygen Inc
Corning
容量
2.0 mL