一般說明
Generator probes for Benchmark′s handheld homogenizer, the D1000.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z742489-1EA | 4061835329670 |