材料
blue gloves
blue nitrile
尺寸
S
無菌
non-sterile
特點
CE compliant
disposable
latex free
powder-free
包裝
pack of 50 ea (boxes of 50)
製造商/商標名
KCL 743 - 07
KCL 743
長度 × 厚度
270 mm × 0.2 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Dermatril P is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril P disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril P disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
All Dermatril P disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
特點和優勢
- Anti-static
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- Roughened fingertips for better grip
- Rolled cuff edge for easier donning
- CE compliant
- Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
法律資訊
Dermatril is a trademark of KCL GmbH
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z677361-10X50EA | 4061833069455 |
Z677361-50EA | 4061833069462 |