主要文件
Z609676
Bohlender screw caps
GL 32 thread
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.17
製造商/商標名
Bohlender
存貨情形
not available in USA and Canada
接頭
GL 32 thread
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
法律資訊
Bohlender is a trademark of Bohlender GmbH
历史批次信息供参考:
分析证书(COA)
访问文档库
Global Trade Item Number
货号GTIN
252115-25G
252115-5G4061825945613
Z609676-10EA