製造商/商標名
Oakton
測量範圍
0-50 °C, accuracy: 1 °C (ATC)
pH測量範圍
0.00-14.99 pH unit, accuracy: 0.01 pH unit, resolution: 0.01 pH unit
正在寻找类似产品? 访问 产品对比指南
一般說明
Features toggle readout of pH and temperature.
Waterproof with unique double junction electrode design that is ideal for most applications including tris buffers, organics, solutions with heavy metals, sulfides, and dirty water.
- Push-button three-point calibration, 4.01, 7.00, 10.00
- Double-junction gel pH electrode
- Powered by four 1.5V alkaline batteries
法律資訊
pHTestr is a registered trademark of Oakton
Global Trade Item Number
货号 | GTIN |
---|---|
Z527831-1EA | 4061833353493 |