主要文件
Z260215
Sigma-Aldrich
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 2-10 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
特點
subdivision 0.2 mL
製造商/商標名
Fortuna Graf 101.107-38
體積測量範圍
2-10 mL
容積
2-10 mL
接頭
GL 40
GL 45
正在寻找类似产品? 访问 产品对比指南
一般說明
For dispensing strong acids and aggressive chemicals
Features:
  • PTFE encased dispensing piston with sealing lip at its end
  • FIX adapter made of PTFE
  • Valve block made of PTFE, no metal springs in the valve - no corrosion
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
  • Ergonmic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
Supplied as Optifix® Safety dispenser, 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
法律資訊
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
历史批次信息供参考:
分析证书(COA)