主要文件
Z136808
Ace Friedrichs condenser
joint: ST/NS 24/40
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
接合處
joint: ST/NS24/40
製造商/商標名
Ace Glass 597410
相关类别
一般說明
Top of condenser is modified with Ace-Thred so that inlet is removable and replaceable. Molded spiral condensing surface. Includes body, inlet tube, and bushing.
历史批次信息供参考:
分析证书(COA)