主要文件
Z100862
Stainless steel 304 syringe needle
gauge 18, L 24 in.
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122002
eCl@ss:
32070500
NACRES:
NB.03
描述
Standard Hub with 304 Stainless Steel Tubing
包裝
pack of 1 ea
製造商/商標名
Cadence Science
長度
24 in.
測量計
18
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)
访问文档库
Global Trade Item Number
货号GTIN
Z100862-1EA4061832529455