SYNF901425
别名:
Synthware F901425, modified Schlenk reaction flask
1 of 10
Sigma-Aldrich
Z515981
Schlenk reaction and storage tube
Z515752
Schlenk Kjeldahl reaction and storage flask
Z515876
Aldrich® solvent storage flask
Z515701
Z515922
Z414484
Aldrich® single-neck round-bottom flask
Z515671
Z515884
SYNF904500
Synthware™ pear-shaped Schlenk flask with stopcock
Z515906
PTFE stopcockborosilicate glass flaskpear-shaped single-neck
joint: ST/NS14/20
pkg of 1 ea
Synthware F901425
25 mL
正在寻找类似产品? 访问 产品对比指南
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.
如需帮助,请联系 客户支持
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 9
HAM203220
Hamilton® SaltLine reagent syringe
SYNA264100
Synthware™ Kjeldahl distilling trap
Z173231
Reaction and storage flask, Kjeldahl, with glass stopcock on side-arm
HAM203230
Z173061
Solvent storage flask with glass stopcock on side-arm
Z220264
Reaction flask with glass stopcock on side-arm
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z648299
微量称重勺和小铲套装