主要文件
SLW3120/14M
Pyrex® Sintered glass filter disc
nominal diameter 120 mm, porosity grade 4
别名:
Pyrex, SciLabware 3120/14M
登录查看公司和协议定价
About This Item
分類程式碼代碼:
40161507
NACRES:
NB.24
材料
Pyrex membrane
無菌
non-sterile
特點
grade 4
製造商/商標名
SciLabware 3120/14M
直徑
120 mm
標稱直徑
120 mm
孔徑
grade 4 porosity
正在寻找类似产品? 访问 产品对比指南
一般說明
  • Complies with ISO 4793
  • For use by Glassblowers constructing special apparatus
  • Manufactured from Pyrex borosilicate glass for resistance to heat and chemical corrosion
  • Close control of pore size
  • Accurate and consistent disc dimensions
  • High mechanical strength to withstand full vacuum
  • Convenient packaging
法律資訊
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)
访问文档库
Global Trade Item Number
货号GTIN
SLW3120/14M-1EA4061832439853