主要文件
SLW1517/18
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 10000 mL
别名:
Pyrex, SciLabware 1517/18
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.17
容量:
10000 mL
直徑× 高度:
227 mm × 410 mm
材料:
borosilicate glass bottle
round bottle
white graduation
材料
borosilicate glass bottle
round bottle
white graduation
接合處
Joints threaded neck (GL45)
特點
graduations
without cap
with printed trace code
without pouring ring
包裝
pack of 1 ea
製造商/商標名
SciLabware 1517/18
容量
10000 mL
直徑× 高度
227 mm × 410 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
特點和優勢
• Bottles only. Supplied without cap or pouring ring.• Complies with ISO 4796.• Printed with Trace Code for downloadable batch certificate.• Sloping shoulders for smooth pouring action.• White graduations and marking spot.• Manufactured from temperature and chemically resistant borosilicate glass.• Choice of caps and pouring rings available separately.• The 25 ml size bottle features a glass lip and does not require a pouring ring.
法律資訊
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)