主要文件
PL800GN
Plas-Labs neoprene glove box gloves
for 1 piece glove set, size 9 (for 8 in. ports)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
46181504
NACRES:
NB.35
材料
neoprene
尺寸
9 (for 8 in. ports)
特點
reusable
包裝
pack of 1 pair
製造商/商標名
Plas-Labs 800GN
正在寻找类似产品? 访问 产品对比指南
一般說明
Plas-Labs black neoprene rubber gloves for chemical resistance
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
历史批次信息供参考:
分析证书(COA)