主要文件
PL800GA-BUT-9
Plas-Labs replacement butyl gloves
for 2 piece glove set, size 9 (for 8 in. ports)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
46181504
NACRES:
HA.03
材料
butyl rubber
尺寸
9 (for 8 in. ports)
特點
reusable
for use with glove box sleeves
包裝
pack of 1 pair
製造商/商標名
Plas-Labs 800GA-BUT-9
正在寻找类似产品? 访问 产品对比指南
一般說明
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
历史批次信息供参考:
分析证书(COA)