主要文件
NORTCM200
Bruker tapered PTFE caps
for NORS205001
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
SB.32
特點
for NORS205001
包裝
pack of 5 ea
製造商/商標名
Norell TCM200
正在寻找类似产品? 访问 产品对比指南
一般說明
Bruker Match tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
法律資訊
Match is a trademark of Bruker Biospin Corp.
历史批次信息供参考:
分析证书(COA)