主要文件
MT51344830
Mettler-Toledo perfectION combination electrode
Measurement range 10-6-0.1 mol/L & 0.2-20700 mg/L
登录查看公司和协议定价
About This Item
分類程式碼代碼:
39121436
NACRES:
NB.79
材料
epoxy shaft
glass shaft (solid state)
描述
Lemo connector
製造商/商標名
Mettler-Toledo 51344830
電纜尺寸
1.2 m
正在寻找类似产品? 访问 产品对比指南
一般說明
comb Pb Lemo
Mettler Toledo perfectION ISE Combined ion-selective electrode means ISE measurement has never been so simple!
The new Click & Clear junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
  • Sensor with attached cable and BNC connector
  • Reference electrolyte (60 mL)
  • Operating instructions
    To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
法律資訊
Click & Clear is a trademark of Mettler-Toledo, Inc.
perfectION is a trademark of Mettler-Toledo, Inc.
从最新的版本中选择一种:
分析证书(COA)