主要文件
MT30207958
Mettler Toledo Seven2Go DO meter S4
Meter Only, polarographic
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115611
NACRES:
NB.61
描述
connector BNC/RCA
pH/ion/ORP meters
特點
Meter Only, polarographic
製造商/商標名
Mettler-Toledo 30207958
dissolved oxygen measuring range
0.00-99.9 mg/L (ppm)
壓力測量範圍
500-1100 (mbar)
saturation measuring range
0.0-600% (DO)
溫度測量範圍
0-60 °C
電池電量
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
正在寻找类似产品? 访问 产品对比指南
一般說明
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
  • Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
  • Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
  • Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
  • Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
特點和優勢
Includes: Meter, USB cable, electrode clip, base stand, instructions, quick guide, test certificate, declaration of conformity, batteries
法律資訊
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.
历史批次信息供参考:
分析证书(COA)