主要文件
BR144635
BRAND® BISTABIL®磨砂玻璃塞,锥形接头
borosilicate glass 3.3, joint: ST/NS 29/32, (hollow, hexagonal grip)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
材料
borosilicate glass 3.3
接合處
joint: ST/NS29/32
製造商/商標名
BRAND 144635
正在寻找类似产品? 访问 产品对比指南
一般說明
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
历史批次信息供参考:
分析证书(COA)