Key Documents
BAF379200000
SP Bel-Art® Plastic Forceps
polypropylene, L 5 in. (127 mm)
别名:
plastic forcep, reusable forceps
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122800
NACRES:
NB.43
材料
polypropylene
包裝
pkg of 6 ea
製造商/商標名
Bel-Art F37920-0000
長度
5 in. (127 mm)
一般說明
These polypropylene plastic forceps have serrated tips to ensure a firm grasp.
特點和優勢
  • Strong and durable
  • Steam autoclavable up to 121ºC (250ºF)
法律資訊
Bel-Art is a registered trademark of SP Scienceware, a division of SP
从最新的版本中选择一种:
分析证书(COA)