主要文件
ACE585320
Ace-Safe® tubing connector, UHDPE
#15 to 1/4 in. I.D. tubing
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41106514
NACRES:
NB.51
包裝
pack of 1 ea
製造商/商標名
Ace Glass 5853-20
參數
-20-110 °C temp. limit
正在寻找类似产品? 访问 产品对比指南
一般說明
Tubing connector, used to connect flexible tubing (1/4 in., 3/8 in., 1/2 in., 3/4 in., 1 in. I.D.) to #7, #11, #15 or #25 Ace-Thred for easy, safe connect/disconnect. Nylon bushing slides over serrated end and secures PP connector in thread with silicone O-ring in front groove to make compression seal. With tubing attached at serrated end, un-thread bushing and connector is removed from thread.
法律資訊
Ace-Safe is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)