Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702689

Sigma-Aldrich

Anti-RNF40 Antibody, clone 8I7R5, Rabbit Monoclonal

Synonym(s):

BRE1B, RBP95, STARING

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8I7R5, monoclonal

form

liquid

mol wt

130 kDa

species reactivity

mouse, mouse

concentration

1.65 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

RELGRENRRLQDLATQLQEKHHRISLEYSELQDKVTSAETKVLEMETTVEDLQWDIEKLRKREQKLNKHLAEALEQLNSGYYVSGSSSGFQGGQITLSMQKFEMLNAELEENQELANSRMAELEKLQAELQGAVRTNERLKVALRSLPEEVVRETGEYRMLQAQFSLLYNESLQVKTQLDEARGLLLATKNSHLRHIEHME

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF40(9810)

General description

The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2011]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 231-431 of human RNF40 (O75150).

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service