Z180084
Ace large chromatography column
I.D. × L 50 mm × 1,200 mm, capacity 2.35 L
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
for chromatography
manufacturer/tradename
Ace Glass 582058
technique(s)
LPLC: suitable
I.D. × L
50 mm × 1,200 mm
capacity
2.35 L
Looking for similar products? Visit Product Comparison Guide
General description
Rugged borosilicate glass column with #50 internal Ace-Threds on ends to which a wide variety of upper and lower adapters can be fitted. Suitable for ion-exchange or activated-carbon work. Order column adapters separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service