Skip to Content
Merck
All Photos(1)

Key Documents

M5144

Sigma-Aldrich

4-O-Methyl-D-glucurono-D-xylan

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201

biological source

wood (beech wood)

form

powder

Application

4-O-Methyl-D-glucurono-D-xylan has been used in a study to assess ecto-glycanases and metabolic stability of the capsule in Cryptococcus neoformans. It has also been used in a study that investigated soluble chromogenic substrates for the assay of xylanases and glucanases.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danka Maceková et al.
Journal of basic microbiology, 46(6), 470-479 (2006-12-02)
We have identified a number of ecto-glycanases (glycosylhydrolases) associated with the capsule and/or the cell wall of Cryptococcus neoformans. The enzyme activities detected included alpha-mannosidase, alpha-, and beta -glucosidase, alpha-, and beta-galactosidase, beta-xylosidase, beta-glucuronidase, and endo-beta-1,3-glucanase. Small portions of the
Thu V Vuong et al.
Biotechnology for biofuels, 13, 51-51 (2020-03-20)
Dicarboxylic acids offer several applications in detergent builder and biopolymer fields. One of these acids, 4-O-methyl d-glucaric acid, could potentially be produced from glucuronoxylans, which are a comparatively underused fraction of wood and agricultural biorefineries. Accordingly, an enzymatic pathway was
P Biely et al.
Analytical biochemistry, 144(1), 142-146 (1985-01-01)
New soluble chromogenic substrates were prepared for specific and rapid assays of endo-1,4-beta-xylanases and endo-1,4-beta-glucanases. A soluble beechwood 4-O-methyl-D-glucurono-D-xylan was dyed with Remazol brilliant blue R, and hydroxyethylcellulose was coupled to Ostazin brilliant red H-3B. The assays are based on
Junpei Zhou et al.
Folia microbiologica, 59(5), 423-431 (2014-04-15)
A glycosyl hydrolase family 10 endoxylanase from Bacillus sp. HJ14 was grouped in a separated cluster with another six Bacillus endoxylanases which have not been characterized. These Bacillus endoxylanases showed less than 52% amino acid sequence identity with other endoxylanases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service