Skip to Content
Merck
All Photos(1)

Key Documents

ABN480

Sigma-Aldrich

Anti-SELK Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Selenoprotein K, HSPC030, HSPC297

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SELK(58515)

General description

Selenoprotein K (SELK) is a selenoprotein that is localized at the endoplasmic reticulum, and highly expressed in the heart. SELK functions as an antioxidant that can bind to proteins such as the ER-associated degradation (ERAD) components p97 ATPase, Derlin-1 and -2, and SelS as well as proteins in the oligosaccharyltransferase complex. SELK may be involved in the Derlin-dependent ERAD of glycosylated misfolded proteins, is thought to be important in Ca2+ flux in immune cells, and is cleaved by m-calpain in resting macrophages.

Immunogen

KLH-conjugated linear peptide corresponding to human SELK.

Application

Anti-SELK Antibody is a highly specific rabbit polyclonal antibody, that targets Selenoprotein K & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in A20 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected SELK in 15 µg of A20 cell lysate.

Target description

~10 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A20 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service