Skip to Content
Merck
L-Tryptophan Vetec™, reagent grade, ≥98%

L-Tryptophan

L-Tryptophan Vetec™, reagent grade, ≥98%
Synonym(s):
(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid, L-Tryptophan, (S)-(-)-Tryptophan
Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Optical Isomer
Amino Acid
Technique