Skip to Content
Merck
All Photos(2)

Key Documents

V900459

Sigma-Aldrich

L-Histidine

≥99%, Vetec

Synonym(s):

(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Product Name

L-Histidine, Vetec, reagent grade, ≥99%

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

color

white to off-white

mp

282 °C (dec.) (lit.)

application(s)

detection

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urszula Kosinska Eriksson et al.
Science (New York, N.Y.), 340(6138), 1346-1349 (2013-06-15)
Aquaporins are membrane channels that facilitate the flow of water across biological membranes. Two conserved regions are central for selective function: the dual asparagine-proline-alanine (NPA) aquaporin signature motif and the aromatic and arginine selectivity filter (SF). Here, we present the
Camilla Hofström et al.
Journal of medicinal chemistry, 56(12), 4966-4974 (2013-05-23)
Engineered affibody molecules can be used for high contrast in vivo molecular imaging. Extending a recombinantly produced HER2 binding affibody molecule with a hexa-histidine tag allows for convenient purification by immobilized metal-ion affinity chromatography and labeling with [(99m)Tc(CO)3](+) but increases
Pawel Swietach et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), E2064-E2073 (2013-05-17)
Ca(2+) signaling regulates cell function. This is subject to modulation by H(+) ions that are universal end-products of metabolism. Due to slow diffusion and common buffers, changes in cytoplasmic [Ca(2+)] ([Ca(2+)]i) or [H(+)] ([H(+)]i) can become compartmentalized, leading potentially to
H M Golder et al.
Journal of dairy science, 97(8), 5131-5150 (2014-06-03)
Ruminal bacterial community composition (BCC) and its associations with ruminal fermentation measures were studied in dairy heifers challenged with combinations of grain, fructose, and histidine in a partial factorial study. Holstein-Friesian heifers (n=30) were randomly allocated to 5 triticale grain-based
Smriti Kala et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 6022-6027 (2014-04-09)
The genome packaging reactions of tailed bacteriophages and herpes viruses require the activity of a terminase enzyme, which is comprised of large and small subunits. Phage genomes are replicated as linear concatemers composed of multiple copies of the genome joined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service