Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200120

Sigma-Aldrich

Anti-Ephrin-B2 antibody ,Mouse monoclonal

clone EFR-163M, purified from hybridoma cell culture

Synonym(s):

Anti-EFNB2, Anti-EPH-related receptor tyrosine kinase ligand 5, Anti-EPLG5 ligand of EPH-related kinase 5, Anti-HTK ligand, Anti-HTKL, Anti-LERK5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

EFR-163M, monoclonal

form

buffered aqueous solution

mol wt

antigen ~56 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells overexpressing a human Ephrin-B2 fusion protein

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFNB2(1948)

General description

Eph family of tyrosine kinase receptors mediates cell-cell attachment and aggregation by binding to specific ligands, ephrins. The Eph family consists of 14 structurally related receptors that are either membrane linked or are transmembrane with overlapping affinity for ligands. The transmembrane receptor Ephrin-B2 signals through its cognate receptor EphB4. The signalling via EphB4 receptor ensures is crucial in the initial stages of vasculogenesis in marking specific arterial and venous domains. Ephrin-B2 is also essential in the development stages for neural crest formation, angiogenesis, vascular cell assembly, migration and synapse development in central nervous system
Anti-Ephrin-B2 recognizes human Ephrin-B2 protein (~56 kDa).
Ephrin-B2 is a member of the ephrin-B (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases. Ephrins are divided into ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage and the ephrin-B (EFNB) class, which are transmembrane proteins. Ephrin-B2 binds the tyrosine kinase receptors EphB4 and EphA3 and is expressed by arteries whereas its receptor tyrosine kinase EphB4 is expressed by veins.

Immunogen

human Ephrin-B2 recombinant protein. The human protein shares 97% homology with the mouse protein.

Application

Anti-Ephrin-B2 antibody may be used for immunoblotting at a working concentration of 0.5-1.0 μg/ml using whole cell lysates of HEK-293T cells over-expressing human Ephrin-B2. It is also suitable for immunohistochemistry of formalin-fixed, paraffin-embedded human skin sections.

Biochem/physiol Actions

Ephrin-B2 and EphB4 are essential for proper development of the cardiovascular system. It is implicated in regulating erythropoiesis. Mutations in these genes are linked to embryonic lethality, defects in the angiogenic remodeling of the peripheral vasculature and defective myocardial trabeculation in the heart.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functions of ephrin/Eph interactions in the development of the nervous system: emphasis on the hippocampal system
Martinez A and Soriano E
Brain Research Reviews, 49(2), 211-226 (2005)
Regulation of angiogenesis by Eph-ephrin interactions
Kuijper S, et al.
Trends in Cardiovascular Medicine, 17(5), 145-151 (2007)
Ephrin-B2 is a candidate ligand for the Eph receptor, EphB6
Munthe E, et al.
Febs Letters, 466(1), 169-174 (2000)
Regulation of vasculogenesis and angiogenesis by EphB/ephrin-B2 signaling between endothelial cells and surrounding mesenchymal cells
Oike Y, et al.
Blood, 100(4), 1326-1333 (2002)
A role of EphB4 receptor and its ligand, ephrin-B2, in erythropoiesis
Suenobu S, et al.
Biochemical and Biophysical Research Communications, 293(3), 1124-1131 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service