Skip to Content
Merck
All Photos(1)

Key Documents

MXQSEAL01

Millipore

Milliflex® Quantum Membrane Transfer Tool Seals

Spare part - For use with Milliflex® Quantum system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.24

manufacturer/tradename

Milliflex®

input

sample(s) (in process)
sample(s) (final product)
water (final product)
sample(s) (raw material)

application(s)

pharmaceutical

shipped in

ambient

General description

Milliflex® Quantum membrane transfer tool seal is an additional accessory for Milliflex® Quantum Rapid Detection System. Milliflex® Quantum System is a rapid fluorescence-based technology for quantitative detection of contaminants over a broad range of filterable matrices proving as a fast and reliable alternative for the detection of microorganisms. The membrane transfer tool seal is used to seal the groove of the tool and aids in efficient attachment of the Milliflex® PLUS Pump head.

Application

Milliflex® Quantum membrane transfer tool seal is an additional accessory for Milliflex® Quantum Rapid Detection System to ensure proper sealing of the tool to the pump head. It finds application in bioburden testing and water monitoring of beer, bottled water, cosmetics, and Pharmaceutical Products Analysis

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service