Skip to Content
Merck
All Photos(1)

Key Documents

650692

Sigma-Aldrich

Cytidine-13C9,15N3 5′-monophosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

CMP-13C9,15N3 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H1215N3Na2O8P
Molecular Weight:
379.07
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom %

Quality Level

Assay

≥95% (CP)

form

solid

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+12

storage temp.

−20°C

SMILES string

[15NH2][13C]([13CH]=[13CH][15N]1[13C@]2([H])O[13C@@]([13CH2]OP(O[Na])(O[Na])=O)([H])[13C@](O)([H])[13C@]([H])2O)=[15N][13C]1=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service