184659
Cellulose, cyanoethylated
extent of labeling: ~2.6 mol cyanoethylation per mol cellulose (D.S.)
Synonym(s):
Cellulose cyanoethyl ether, Cyanoethyl cellulose
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C36H46N8O11
CAS Number:
Molecular Weight:
766.80
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
shavings
Quality Level
extent of labeling
~2.6 mol cyanoethylation per mol cellulose (D.S.)
transition temp
Tg 180 °C
SMILES string
N#CCCOCCC#N
InChI
1S/C6H8N2O/c7-3-1-5-9-6-2-4-8/h1-2,5-6H2
InChI key
BCGCCTGNWPKXJL-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service