Skip to Content
Merck
All Photos(1)

Key Documents

45922

Supelco

4-Methyl-m-phenylenediamine

analytical standard

Synonym(s):

2,4-Diaminotoluene, 2,4-Toluenediamine, 4-Methyl-1,3-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein:
2205839
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

283-285 °C (lit.)

mp

97-99 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(N)cc1N

InChI

1S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4H,8-9H2,1H3

InChI key

VOZKAJLKRJDJLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-m-phenylenediamine is an industrial intermediate, widely used in the production of industrial dyes and polyurethane foams.

Application

4-Methyl-m-phenylenediamine may be used as an analytical reference standard for the quantification of the analyte in biological fluids using high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-chu Shan et al.
Se pu = Chinese journal of chromatography, 20(4), 289-294 (2003-01-25)
In high performance liquid chromatography, it is necessary to apply multi-composition gradient elution for the separation of complex samples such as environmental and biological samples. Multivariate stepwise gradient elution is one of the most efficient elution modes, because it combines
Pei-Shan Liu et al.
Toxicology, 219(1-3), 167-174 (2005-12-13)
Toluene diisocyanate (TDI) is widely used as a chemical intermediate in the production of polyurethane. TDI-induced asthma is related to its disturbance of acetylcholine activity in most affected workers, but the relevant mechanisms are unclear. Toluene diamine (TDA) is the
Astrid Rohrbeck et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(1), 31-41 (2010-08-18)
Information on the carcinogenic potential of chemicals is primarily available for High Production Volume (HPV) products. Because of the limited knowledge gain from routine cancer bioassays and the fact that HPV chemicals are tested only, there is the need for
2,4-Diaminotoluene.
Report on carcinogens : carcinogen profiles, 10, 78-79 (2004-08-25)
Toshiki Nozaka et al.
Environmental sciences : an international journal of environmental physiology and toxicology, 14 Suppl, 1-8 (2008-07-24)
The subacute toxicity of toluene-2,4-diamine (2,4-TDA) on marine fish was investigated in laboratory toxicity tests using red sea bream, Pagrus major. The fish were exposed to subacute concentrations of 2,4-TDA (measured concentrations: 0.0628, 0.234 and 0.878 mg/L) during the 14-day

Protocols

GC Analysis of Anilines on Equity®-5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service