Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300390

Sigma-Aldrich

Anti-CHEK1 (Ab-317) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CHK1 antibody produced in rabbit, Anti-CHK1 checkpoint homolog (S. pombe) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~56 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-S-S-Q-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHEK1(1111)

Immunogen

Peptide sequence around aa. 315-319 (S-S-S-Q-P), according to the protein CHEK1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Chek1 is a protein kinase that inhibits mitotic entry after DNA damage, required for the DNA damage checkpoint and is strongly similar to murine Chek1. Checkpoint pathways control the order and timing of cell cycle transitions and ensure that critical events, such as DNA replication and chromosome segregation, are completed with high fidelity. The mouse and human proteins share 90% sequence identity through the protein kinase domains. The sequence of the 476-amino acid human Chek1 protein is 29%, 40%, and 44% identical to those of the fission yeast Chek1, C. elegans Chek1, and Drosophila 'grapes' (Grp) proteins, respectively. Chek1 is expressed ubiquitously as an approximately 2.4-kb mRNA, with the most abundant expression in thymus, testis, small intestine, and colon. The protein has altered mobility when isolated from cells treated with ionizing radiation, indicating that Chek1 is modified in response to DNA damage. In vitro, Chek1 directly phosphorylates a regulator of CDC2 tyrosine phosphorylation, CDC25C. In response to DNA damage, Chek1 phosphorylates and inhibits CDC25C, thus preventing activation of the CDC2-Cyclin-B complex and mitotic entry.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin Xie et al.
Medical oncology (Northwood, London, England), 31(3), 844-844 (2014-01-28)
Checkpoint kinase 1 (CHEK1) is an evolutionarily conserved Ser/Thr kinase, which mediates cell-cycle arrest after DNA damage, and we previously reported that CHEK1 was overexpressed and associated with poor prognosis in hepatocellular carcinoma (HCC), indicating it was oncogenic gene. In
Judy Yan et al.
Experimental cell research, 328(1), 132-142 (2014-08-26)
Despite the development of chemoresistance as a major concern in prostate cancer therapy, the underlying mechanisms remain elusive. In this report, we demonstrate that DU145-derived prostate cancer stem cells (PCSCs) progress slowly with more cells accumulating in the G1 phase
Eui Young So et al.
Cancer biology & therapy, 15(7), 906-910 (2014-04-24)
The bone marrow (BM) is one of the organs that is sensitive to acute exposure of ionizing radiation (IR); however, the mechanism of its high sensitivity to IR remains to be elucidated. BM is differentiated into dendritic cells (DC) with
Hiroyasu Sakai et al.
Cell cycle (Georgetown, Tex.), 13(6), 1015-1029 (2014-02-21)
Wip1 (protein phosphatase Mg(2+)/Mn(2+)-dependent 1D, Ppm1d) is a nuclear serine/threonine protein phosphatase that is induced by p53 following the activation of DNA damage response (DDR) signaling. Ppm1d(-/-) mouse embryonic fibroblasts (MEFs) exhibit premature senescence under conventional culture conditions; however, little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service