Skip to Content
Merck
All Photos(1)

Key Documents

8.20046

Sigma-Aldrich

4-Ethyltoluene

for synthesis

Synonym(s):

4-Ethyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
120.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
210-761-2
form:
liquid
Assay:
≥96.0% (GC)

Quality Level

Assay

≥96.0% (GC)

form

liquid

autoignition temp.

475 °C

potency

4850 mg/kg LD50, oral (Rat)

expl. lim.

1.2-7 % (v/v)

kinematic viscosity

0.812 cSt(20 °C)

bp

160-162 °C/1013 hPa

mp

-62 °C

transition temp

flash point 43 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H12/c1-3-9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

JRLPEMVDPFPYPJ-UHFFFAOYSA-N

Application

4-Ethyltoluene for synthesis. CAS 622-96-8, chemical formula 4-(C₂H₅)C₆H₄CH₃.

Specifications

Saturation Concentration in Air: 16 g/m3 (20 °C) Air

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.86 - 0.88
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service