Skip to Content
Merck
All Photos(2)

Key Documents

8.05802

Sigma-Aldrich

2-(Methylamino)ethanol

for synthesis

Synonym(s):

2-(Methylamino)ethanol, N-Methylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO
CAS Number:
Molecular Weight:
75.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-710-0
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

350 °C

potency

1391 mg/kg LD50, oral (Rat)
1006 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-17.7 % (v/v)

refractive index

n20/589 1.4387

pH

13.6 (20 °C, 100 g/L in H2O)

bp

159 °C/1013 hPa

mp

-3 °C

transition temp

flash point 74 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3

InChI key

OPKOKAMJFNKNAS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.938 - 0.940
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service