1.09719
n-Pentane
reference substance for gas chromatography
Synonym(s):
Pentane
About This Item
Recommended Products
vapor density
2.48 (vs air)
Quality Level
Assay
≥99.7% (GC)
form
liquid
autoignition temp.
500 °F
potency
>2000 mg/kg LD50, oral (Rat)
expl. lim.
~8.3 %
technique(s)
gas chromatography (GC): suitable
refractive index
n20/D 1.358 (lit.)
bp
35-36 °C (lit.)
mp
−130 °C (lit.)
transition temp
flash point -48 °C (refers to pure substance)
solubility
0.4 g/L
density
0.626 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
CCCCC
InChI
1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
InChI key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Identity (IR): conforms
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-40.0 °F
Flash Point(C)
-40 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service