Skip to Content
Merck
All Photos(1)

Key Documents

1.07251

Supelco

Phenylhydrazine

GR for analysis

Synonym(s):

Phenylhydrazine, Hydrazinobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
MDL number:
UNSPSC Code:
12352127
EC Index Number:
202-873-5

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

195 °C

potency

188 mg/kg LD50, oral (Rat)

expl. lim.

1.1 % (v/v)

bp

244 °C/1013 hPa

mp

19.6 °C

transition temp

flash point 88 °C

solubility

145 g/L

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H

InChI key

JOVOSQBPPZZESK-UHFFFAOYSA-N

General description

GR for analysis

Application

Phenylhydrazine GR for analysis. CAS 100-63-0, EC Number 202-873-5, chemical formula C₆H₅NHNH₂.

Analysis Note

Assay (HClO₄)≥ 97.0 %Density (d 20 °C/ 4 °C)1.096 - 1.099Identity (IR)passes testDue to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service