Polymer Synthesis Techniques
Polymers are a class of materials comprising small repeating chemical units. The building block or the repeating unit in the polymer is called a monomer. Polymerization, or polymer synthesis, is a chemical reaction in which monomers are joined together by covalent bonding to form polymer structures. The length of the polymer chain is characterized by the number of repeating units in the chain, which is referred to as degree of polymerization (DP). The molecular weight of the polymer is the product of the molecular weight of the repeating unit and the DP. The basic properties inherent to polymers mainly depend on the molecular weight, structure (linear or branched), and DP.
Featured Categories
We provide reliable polymerization tools to initiate and manipulate a chemical reaction of monomers to form polymer chains or networks with specific properties for diverse applications.
Discover our selection of polyethylene glycol (PEGs) and PEG derivatives in a wide range of molecular weights for all your PEGylation needs and applications.
We offer an extensive range of natural and synthetic biomedical polymers with advanced properties suitable for all your biomedical applications.
Explore our portfolio of versatile hydrophobic polymers to support your adhesion, coating, fiber, film, engineered plastic and biomedical applications.
Based on the types of chemical reaction involved, polymerizations are grouped into two types, known as condensation and addition polymerization. In condensation or step-growth polymerization, a condensation reaction takes place between two different bi-functional or tri-functional monomers to produce polymers, whereby small molecules, often water, are eliminated from the reaction. In addition polymerization, or chain-growth polymerization, the growth of the polymer occurs by adding monomers to active sites on the polymer chain with regeneration of the active sites at the end of each growth step. An initiator is required to produce the initiator species with reactive centers. The reactive center can be a free radical, cation, anion, or organometallic complex.
The characteristics of polymers are highly dependent on monomer types and the polymerization method used for the synthesis. Living radical polymerization, also referred to as controlled radical polymerization (CRP), enables better control over the polymer molecular weight, molecular weight distribution, functionality, and composition. CRP can be utilized with a wide range of vinyl monomers to build polymers with diverse properties for different applications.
There are three fundamental CRP techniques:
- Atom transfer radical polymerization (ATRP)
- Reversible addition/fragmentation chain transfer (RAFT) polymerization
- Nitroxide-mediated polymerization (NMP)
ATRP is well-suited for surface modification applications which rely on tailored hydrophilicity, adhesive properties, or nanoparticle functionalization. Polymers derived from both RAFT and ATRP can be modified post-polymerization and are used extensively in the biomedical field for drug delivery and tissue engineering applications. Block copolymers for biomedical applications, are commonly developed from RAFT or ATRP. Block copolymers, developed from NMP, are essential for pigment dispersion, memory devices, and composite manufacturing.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Find reversible addition–fragmentation chain transfer(RAFT) polymerization process, classes of RAFT agents & applications.
- Literature values for the glass transition temperature, (Tg), and melting temperature, (Tm), are given for the more common homopolymers.
- Refractive index (RI or n) is of critical importance for photonics applications such as optical waveguides and ophthalmic devices. Based on their unique refractive index characteristics and good optical clarity, polymeric materials are also used as anti-reflective coatings for solar cells, displays and contact lenses.
- MALDI-TOF MS is a tool for confirming end group identity & transformations, verifying successful & complete conjugation. Functionalized polymers enable a quick & efficient route for synthesis of various polymer conjugates for many applications if apt end group is present.
- Atom transfer radical polymerization (ATRP) has emerged as one of the most successful synthetic techniques for the preparation of polymers with predetermined molecular weights, narrow molecular weight distributions, and high degrees of chain end functionalities.
- See All (55)
Related Protocols
- An article about the typical procedures for polymerizing via ATRP, which demonstrates that in the following two procedures describe two ATRP polymerization reactions as performed by Prof. Dave Hadddleton′s research group at the University of Warwick.
- Monodisperse, surfactant-free polymer spheres for use as colloidal crystal templates can be easily obtained in reasonably large quantities. Typical synthesis methods for poly(methyl methacrylate) (PMMA) and poly(styrene) (PS) by emulsifier free emulsion polymerization are described below and yield spheres several hundred nanometers in diameter.
- RAFT (Reversible Addition-Fragmentation chain Transfer) is a form of living radical polymerization involving conventional free radical polymerization of a substituted monomer in the presence of a suitable chain transfer (RAFT) reagent.
- We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.
- We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.
- See All (6)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email custserv@sial.com
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ