- Mapping of the gene coding for the human liver/bone/kidney isozyme of alkaline phosphatase to chromosome 1.
Mapping of the gene coding for the human liver/bone/kidney isozyme of alkaline phosphatase to chromosome 1.
Annals of human genetics (1986-07-01)
D M Swallow, S Povey, M Parkar, P W Andrews, H Harris, B Pym, P Goodfellow
PMID3446011
ABSTRACT
The gene coding for the liver/bone/kidney isozyme of alkaline phosphatase, ALPL, has been mapped to human chromosome 1 using a monoclonal antibody TRA-2-54/2J and electrophoretic analysis to distinguish between the human and rodent isozymes in human/rodent somatic cell hybrids.