Merck
  • para-acetamidobenzoylglutamate is a suitable indicator of folate catabolism in rats.
para-acetamidobenzoylglutamate is a suitable indicator of folate catabolism in rats.
The Journal of nutrition (1995-10-01)
F L Geoghegan, J M McPartlin, D G Weir, J M Scott
ABSTRACT
The amount of biologically active folate excreted in the urine corresponds to a small fraction of the recommended dietary allowance, suggesting that a large amount of folate is excreted as its catabolites. A strategy of assessing folate requirement by measuring the daily urinary levels of products of folate catabolism depends on the demonstration of an exclusive mechanism of breakdown as well as a suitable marker of the catabolic process. Rats were given [3H] and [14C]folic acid by gastric intubation daily for 10 d to simulate normal dietary intake of the vitamin. Total urine was collected throughout this period as well as for the following 10 d. Reverse-phase HPLC of the radiolabeled urinary products revealed the presence of a variety of intact folates as well as products of C9-N10 scission of the folate molecule, pteridines, para-aminobenzoylglutamate and para-acetamidobenzoylglutamate. We detected no other N10-containing catabolites, nor did we find the oxidized folate derivative '4 alpha-hydroxy-5-methyltetrahydrofolate'. Of all the urinary folate metabolites, only para-acetamidobenzoylglutmate persisted at high levels up to 10 d after radiolabel treatment was withdrawn. We conclude that folate catabolism occurs exclusively through C9-N10 cleavage and that measurement of urinary para-acetamidobenzoylglutmate provides a suitable indicator of daily folate turnover.
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%