Merck
Induction of premature mitosis in S-blocked onion cells.
Cell biology international (2000-06-30)
R Sen, S Ghosh
ABSTRACT
Onion root-tip cells were blocked at S-phase by treating them with 5-aminouracil (5AU). These cells were then further treated with caffeine/2-aminopurine (Caf/2AP) or a combination of both in the presence of 5AU. These tyrosine kinase inhibitors were able to induce premature mitosis in the S-blocked cells as evident from the breaks and gaps in the metaphase chromosomes and the presence of laggards and fragments in anaphase. Immunofluorescence showed normal spindle formation in these cells. Immunoblotting of cyclin B revealed that the level of cyclin B was slightly higher in the recovered and treated samples than the S-blocked one. The level of p(34)was found to be almost equal in all three samples as expected. We failed to observe any significant difference in the level of p(34)containing phosphorylated tyrosine. Such premature induction of mitosis by the purine derivatives has also been reported in BHK cells. However, those cells failed to progress through mitosis. A comparative analysis indicates that the plant cells and the animal cells, perhaps, follow identical pathway for the initiation of mitosis. The possible causes for differential behaviour in mitotic progression in these cells have been discussed.
Sigma-Aldrich
5-Aminouracil, 98%