Merck
  • Xanthogranulomatous oophoritis secondary to talcum powder. Case report and review of the literature.
Xanthogranulomatous oophoritis secondary to talcum powder. Case report and review of the literature.
Le Journal medical libanais. The Lebanese medical journal (2012-12-04)
Camil J Chouairy, Elias A Hajal, Yasmine A Nehme
ABSTRACT
Xanthogranulomatous inflammation of the ovary (xanthogranulomatous oophoritis) is a rare condition, usually representing a complication of tubo-ovarian abscess following pelvic inflammatory disease (PID). We report a case of xanthogranulomatous oophoritis that occurred in a 31-year-old Lebanese lady, eight years following an open appendectomy as a reaction to talcum powder present on surgical gloves.