Merck
Mast cell death induced by 24(S),25-epoxycholesterol.
Experimental cell research (2010-09-22)
Makiko Fukunaga, Satoshi Nunomura, Shigeru Nishida, Kaori Endo, Yasuhiro Gon, Shu Hashimoto, Yuichi Hashimoto, Yoshimichi Okayama, Makoto Makishima, Chisei Ra
ABSTRACT
Mast cell is one of the central effectors in inflammatory responses. Recent studies suggest that a promising therapeutic approach for various inflammatory immune diseases, including rheumatoid arthritis, multiple sclerosis, and type I allergies, is to inhibit mast cell growth and/or survival. Studies also indicate that a balanced lipid metabolism is crucial for regulating the life span of cells. Oxysterol is a well-known regulator of lipid metabolism and has diverse functions, such as inhibition of the mevalonate isoprenoid pathway, efflux of free cholesterols, and synthesis of cholesterol esters. Here, we show that 24(S),25-epoxycholesterol, a representative endogenous oxysterol, induces apoptosis in bone marrow-derived murine mast cells. Furthermore, we have revealed, for the first time, that the accumulation of neutral lipids catalyzed by acyl-CoA:cholesterol acyltransferase in the cells was involved in induction of mast cell apoptosis. Our present findings confer new insights into the roles of lipid metabolism during oxysterol-mediated mast cell apoptosis.
Sigma-Aldrich
Sandoz 58-035, >98% (HPLC), powder